| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012952.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012952.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012952.hg.1 |
TC09000986.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166858.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166858.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166858.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166858.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166858.hg.1 |
TC0900009774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
ENST00000425633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009774.hg.1 |
NR_026679.1 |
Refseq |