Search
Search

TBL1Y| Gene Symbol: TBL1Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056903.1 | - | 1 | 1553 | 97 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
testis-specific transcript, Y-linked 8B (non-protein coding) |
TTTY8B |
NCRNA00217, TTTY8 |
-437.0kb | Anti-sense | Chr. Y - 6471031 - 6473630 on NCBI Build GRCh38 |
| Downstream |
protein kinase, Y-linked, pseudogene |
PRKY |
PRKXP3, PRKYP |
361.1kb | Sense | Chr. Y - 7273972 - 7381547 on NCBI Build GRCh38 |
Set Membership: |
|