Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03673833_s1
          A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



          Species:
          Human
          Assay Type:
          Non-coding
          Assay ID Hs03673833_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY211382.1 - Chr. 22 - 31329989 - 31330448 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007100.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens 1700

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3437273 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P255027 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 39937 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4535 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3437273 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P255027 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P255027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 8072484 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553623_PM_at - -
          Affymetrix Human Clariom D Assay PSR2200149860.hg.1 TC2200007100.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1553623_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553623_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553623_at2 - -
          Human Genome U95 Set 73764_PM_at - -
          Human Genome U95 Set 73764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 194727 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1768164 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1768164 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1768164 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1768164 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1768164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline