Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03676656_mH
          See other CXCL12 GE Assays ›
          Gene Symbol:
          CXCL12
          Gene Name:
          C-X-C motif chemokine ligand 12
          Gene Aliases:
          IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1
          Chromosome Location:
          Chr.10: 44292088 - 44385097 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03676656_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000609.6 NP_000600.1 1 - 2 157 88
          NM_001033886.2 NP_001029058.1 1 - 2 157 88
          NM_001178134.1 NP_001171605.1 1 - 2 157 88
          NM_001277990.1 NP_001264919.1 1 - 2 157 88
          NM_199168.3 NP_954637.1 1 - 2 157 88
          GenBank mRNA AK292628.1 - 1 - 2 160 88
          AK311814.1 - 1 - 2 155 88
          AU120056.1 - 1 - 2 152 88
          AY644456.1 - 1 - 2 114 88
          AY874118.1 - 1 - 2 144 88
          BC031072.1 - 1 - 2 149 88
          BC039893.1 - 1 - 2 135 5729604 88
          CD172464.1 - 1 - 2 155 88
          CR450283.1 - 1 - 2 65 88
          DA266752.1 - 1 - 2 158 88
          DQ345517.1 - 1 - 2 109 88
          DQ345518.1 - 1 - 2 104 88
          DQ345519.1 - 1 - 2 109 88
          DQ345520.1 - 1 - 2 109 88
          L36033.1 - 1 - 2 144 88
          L36034.1 - 1 - 2 144 88
          U16752.1 - 1 - 2 145 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010468.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001212.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CXCL12

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          positive regulation of endothelial cell proliferation
          cellular calcium ion homeostasis
          chemotaxis
          immune response
          cell adhesion
          signal transduction
          G-protein coupled receptor signaling pathway
          axon guidance
          blood circulation
          regulation of actin polymerization or depolymerization
          adult locomotory behavior
          response to radiation
          response to heat
          response to mechanical stimulus
          response to virus
          telencephalon cell migration
          organ regeneration
          positive regulation of dopamine secretion
          response to peptide hormone
          positive regulation of cell adhesion
          positive regulation of axon extension involved in axon guidance
          positive chemotaxis
          induction of positive chemotaxis
          cell chemotaxis
          chemokine-mediated signaling pathway
          positive regulation of monocyte chemotaxis
          positive regulation of calcium ion import
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of leukocyte tethering or rolling
          negative regulation of leukocyte apoptotic process
          positive regulation of T cell migration
          receptor binding
          chemokine activity
          growth factor activity
          chemoattractant activity
          chemokine receptor binding
          CXCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000343575.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374426.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000374429.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395793.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395794.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000395795.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000488591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 ENST00000496375.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_000609.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001033886.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001178134.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_001277990.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010468.hg.1 NM_199168.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 118740 - -
          ABI Human Genome Survey Microarray Version 2.0 218843 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0