Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03677934_s1
          See other PDCD4-AS1 GE Assays ›
          Gene Symbol:
          PDCD4-AS1
          Gene Name:
          PDCD4 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 110868890 - 110870904 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03677934_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDCD4-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026932.1 1 1950 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011824.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001659.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27869084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478350 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27869093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27869094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27869086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07478347 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 129887 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3645465 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P153642 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18764 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42871 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3645465 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3645465 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478403 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869104 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478394 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869106 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869142 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478403 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07478392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27869134 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P153642 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P153642 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10895450 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P10898172 - -
          Agilent Human 1B Microarray G4111A A_32_P65673 - -
          Agilent Human 1B Microarray G4111A A_32_P869948 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478361 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478352 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478412 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478342 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478389 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478357 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478363 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478387 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478421 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478410 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478354 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478418 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478373 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478348 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07478385 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P10898172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 222307_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC10001659.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10001659.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10001659.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC10001659.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10001659.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021762.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021762.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021762.hg.1 TC10001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10021761.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021761.hg.1 TC10001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10021763.hg.1 NR_026932.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021763.hg.1 TC10001659.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008903.hg.1 ENST00000416249.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008903.hg.1 ENST00000416249.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180973.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180973.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180973.hg.1 TC1000011824.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000011824.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011824.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011824.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011824.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011824.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180971.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180971.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180971.hg.1 TC1000011824.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008903.hg.1 ENST00000416249.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008903.hg.1 ENST00000416249.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180972.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180972.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180972.hg.1 ENST00000420367.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180972.hg.1 NR_026932.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000180972.hg.1 TC1000011824.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 222307_at - -
          Human Genome U133 Plus 2.0 Array 222307_PM_at - -
          Human Genome U133A 2.0 Array 222307_at - -
          Human Genome U133A 2.0 Array 222307_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw555103 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 222307_at11 - -
          Human Genome U95 Set 90695_PM_at - -
          Human Genome U95 Set 90695_at - -
          Human Genome U133 Set 222307_at - -
          Human Genome U133 Set 222307_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043297 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A043297 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3199489 AK093606.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3199489 NR_026932.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline