Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03681096_s1
          See other MAPT-IT1 GE Assays ›
          Gene Symbol:
          MAPT-IT1
          Gene Name:
          MAPT intronic transcript 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 597911 - 600926 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03681096_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAPT-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024560.1 1 2854 90
          GenBank mRNA BF108492.1 - 1 171 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microtubule associated protein tau MAPT DDPAC,
          FTD1,
          FTDP-17,
          MAPTL,
          MSTD,
          MTBT1,
          MTBT2,
          PPND,
          PPP1R103,
          TAU,
          Tau-PHF6,
          tau-40
          Sense Chr. 17 - 596685 - 730446 on NCBI Build GRCh38
          Downstream saitohin STH MAPTIT 100.4kb Sense Chr. 17 - 701369 - 701813 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC17000593.hg.1, TC17_ctg5_hap1000019.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3230439 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4632 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4631 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3797 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3230439 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3230439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914690 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914703 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914709 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340084 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10340086 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30914690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16987997 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340089 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16987983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340081 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340079 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340091 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16987988 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340086 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10340084 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR17007270.hg.1 NR_024560.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17007270.hg.1 TC17000593.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17_ctg5_hap1000229.hg.1 NR_024560.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17_ctg5_hap1000229.hg.1 TC17_ctg5_hap1000019.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700008083.hg.1 ENST00000624111.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700008083.hg.1 ENST00000624111.2 Ensembl
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009617 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009617 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3247621 NR_024560.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3247621 AK091698.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3247621 NR_024560.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3247621 AK091698.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3247621 NR_024560.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3247621 AK091698.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3247621 NR_024560.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3247621 AK091698.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline