Search
Search
| Gene Symbol: API5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006718359.3 | XP_006718422.1 | 14 | 2137 | 72 | |
| XM_017018464.1 | XP_016873953.1 | 15 | 2194 | 72 | ||
| GenBank mRNA | AK021649.1 | - | 1 | 46 | 72 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heterogeneous nuclear ribonucleoprotein K pseudogene 3 | HNRNPKP3 | -70.3kb | Anti-sense | Chr. 11 - 43261504 - 43269369 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 17 | TTC17 | 16.9kb | Sense | Chr. 11 - 43358885 - 43494933 on NCBI Build GRCh38 |
Set Membership: |
|