| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007823.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
ENST00000570167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156973.hg.1 |
TC1600007823.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600156974.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156974.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156974.hg.1 |
ENST00000570241.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156974.hg.1 |
NR_110908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156974.hg.1 |
TC1600007823.hg.1 |
Clariom D TC |