Search
Search
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | J05063.1 | - | 1 | 656 | 88 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | maestro heat like repeat family member 2B | MROH2B | HEATR7B2 | -87.9kb | Sense | Chr. 5 - 40998020 - 41071342 on NCBI Build GRCh38 |
| Downstream | phosphatidylinositol specific phospholipase C X domain containing 3 | PLCXD3 | 146.8kb | Sense | Chr. 5 - 41306946 - 41510628 on NCBI Build GRCh38 |
Set Membership: |
|