Search
Search

| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | U17620.1 | - | 57 | Chr. - - [-] on Build | 87 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
Set Membership: |
|