Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03840016_m1
          See other LOC100289333 GE Assays ›
          Gene Symbol:
          LOC100289333
          Gene Name:
          uncharacterized LOC100289333
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 12195015 - 12238647 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03840016_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100289333
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK023304.1 - 1 - 2 131 99
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AU133744.1 - 1 - 2 131 Chr. 19 - 12195015 - 12237432 [+] on Build GRCh38 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 136 ZNF136 pHZ-20 -5.1kb Sense Chr. 19 - 12163096 - 12189871 on NCBI Build GRCh38
          Downstream zinc finger protein 563 ZNF563 79.2kb Anti-sense Chr. 19 - 12317477 - 12347390 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 75913 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 147211 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 638803 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 853648 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 181613 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 176269 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 649518 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 63738 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 589797 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P20960442 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P20960442 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03424343 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P20960459 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 123286 AB209372.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 123286 BC038582.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 123286 BC050554.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 123286 NM_006116.2 refseq_mrna_human_NM
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 123286 U49928.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 176059 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 84205 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41143491 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960459 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960442 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03424343 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P12985672 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41143497 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960447 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03424337 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960463 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03424342 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960450 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960423 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20960437 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03424347 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03424351 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20960442 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20960450 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41143491 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P20960459 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03424337 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 233775_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011879.hg.1 ENST00000597563.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000415793.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000426044.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000440004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 ENST00000451691.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 NR_104179.1 Refseq
          Affymetrix Human Clariom D Assay TC1900007068.hg.1 NR_104179.1 Refseq
          Human Genome U133 Plus 2.0 Array 233775_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 233775_x_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233775_x_at5 - -
          Human Genome U133 Set 233775_PM_x_at - -
          Human Genome U133 Set 233775_x_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline