| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150664.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150664.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150664.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150664.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150664.hg.1 |
TC0700007302.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700150665.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150665.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150665.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150665.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150665.hg.1 |
TC0700007302.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007302.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150663.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150663.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150663.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150663.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150663.hg.1 |
TC0700007302.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700150666.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150666.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150666.hg.1 |
ENST00000440213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150666.hg.1 |
NR_110832.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150666.hg.1 |
TC0700007302.hg.1 |
Clariom D TC |