| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016239.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016239.hg.1 |
TC10001298.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016240.hg.1 |
ENST00000426785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016240.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10016240.hg.1 |
TC10001298.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000172652.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000172652.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000172652.hg.1 |
TC1000010654.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000172653.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000172653.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000172653.hg.1 |
ENST00000426785.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000172653.hg.1 |
NR_038277.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000172653.hg.1 |
TC1000010654.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1561558_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1561558_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1561558_at1 |
- |
- |