Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03887660_s1
          See other SNU13 GE Assays ›
          Gene Symbol:
          SNU13
          Gene Name:
          small nuclear ribonucleoprotein 13
          Gene Aliases:
          15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1
          Chromosome Location:
          Chr.22: 41673933 - 41690480 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03887660_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNU13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017028811.2 XP_016884300.1 3 699 61
          GenBank mRNA AK124020.1 - 1 954 61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG822290.1 - 1 476 Chr. 22 - 41675038 - 41688328 [-] on Build GRCh38 61
          GenBankmRNA AI288202.1 - 1 137 Chr. 22 - 41687727 - 41688223 [+] on Build GRCh38 61
          GenBankmRNA CB047182.1 - 1 148 Chr. 22 - 41687742 - 41688284 [+] on Build GRCh38 61
          GenBankmRNA AI572838.1 - 1 122 Chr. 22 - 41687742 - 41688169 [+] on Build GRCh38 61
          GenBankmRNA AI271774.1 - 1 145 Chr. 22 - 41687719 - 41688134 [+] on Build GRCh38 61
          GenBankmRNA AI826286.1 - 1 121 Chr. 22 - 41687743 - 41688203 [+] on Build GRCh38 61
          GenBankmRNA AI278228.1 - 1 130 Chr. 22 - 41687742 - 41688142 [+] on Build GRCh38 61
          GenBankmRNA BQ021175.1 - 1 343 Chr. 22 - 41687537 - 41688079 [+] on Build GRCh38 61
          GenBankmRNA AI682289.1 - 1 121 Chr. 22 - 41687743 - 41688166 [+] on Build GRCh38 61
          GenBankmRNA AI805868.1 - 1 124 Chr. 22 - 41687742 - 41688207 [+] on Build GRCh38 61
          GenBankmRNA AI288207.1 - 1 137 Chr. 22 - 41687727 - 41688174 [+] on Build GRCh38 61

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein LLP homolog LOC107985567 -9.5kb Sense Chr. 22 - 41675202 - 41677160 on NCBI Build GRCh38
          Downstream CTA-216E10.6 C22orf46P C22orf46 121.0 Anti-sense Chr. 22 - 41688937 - 41698136 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          box C/D snoRNP assembly
          mRNA processing
          single fertilization
          RNA splicing
          maturation of SSU-rRNA
          ribosome biogenesis
          ribosomal small subunit biogenesis
          RNA binding
          protein binding
          snoRNA binding
          U4 snRNA binding
          U4atac snRNA binding
          U3 snoRNA binding
          box C/D snoRNA binding
          ATPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000215956.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000355257.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000401959.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000402458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000402458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000402458.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000463675.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000463675.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000463675.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000463675.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469028.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000469522.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000488571.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000488571.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 ENST00000488571.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_001003796.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008837.hg.1 NM_005008.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline