Search
Search

GABRB1| Gene Symbol: GABRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007985.1 | XP_016863474.1 | 5 | 9829 | 100 | |
| GenBank mRNA | AK124778.1 | - | 1 | 2951 | 100 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | COMM domain containing 8 | COMMD8 | 14.2kb | Anti-sense | Chr. 4 - 47450793 - 47463702 on NCBI Build GRCh38 |
Set Membership: |
|