Search
Search
| Gene Symbol: CDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170406.1 | NP_001163877.1 | 3 - 4 | 352 | 115 | |
| NM_001170407.1 | NP_001163878.1 | 3 - 4 | 340 | 115 | ||
| NM_001320918.1 | NP_001307847.1 | 3 - 4 | 352 | 115 | ||
| NM_001786.4 | NP_001777.1 | 3 - 4 | 340 | 115 | ||
| NM_033379.4 | NP_203698.1 | 3 - 4 | 340 | 115 | ||
| XM_005270303.3 | XP_005270360.1 | 3 - 4 | 422 | 115 | ||
| GenBank mRNA | AK291939.1 | - | 3 - 4 | 277 | 115 | |
| AK295741.1 | - | 3 - 4 | 318 | 115 | ||
| AW976143.1 | - | 3 - 4 | 255 | 115 | ||
| BC014563.1 | - | 2 - 3 | 211 | 2967728 | 115 | |
| BC107750.1 | - | 3 - 4 | 301 | 6710672 | 115 | |
| BF685285.1 | - | 3 - 4 | 401 | 116 | ||
| BG723076.1 | - | 3 - 4 | 345 | 115 | ||
| BT007004.1 | - | 2 - 3 | 198 | 115 | ||
| CR933728.1 | - | 3 - 4 | 299 | 115 | ||
| D88357.1 | - | 2 - 3 | 225 | 115 | ||
| DA823877.1 | - | 3 - 4 | 353 | 115 | ||
| X05360.1 | - | 3 - 4 | 324 | 115 | ||
| Y00272.1 | - | 3 - 4 | 324 | 115 | ||
Set Membership: |
|