Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04187260_m1
          See other INHBA GE Assays ›
          Gene Symbol:
          INHBA
          Gene Name:
          inhibin beta A subunit
          Gene Aliases:
          EDF, FRP
          Chromosome Location:
          Chr.7: 41685101 - 41710532 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04187260_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: INHBA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002192.2 NP_002183.1 1 - 2 101 117
          GenBank mRNA AF218018.1 - 1 - 2 62 115
          AK290584.1 - 1 - 2 64 117
          AU137531.1 - 1 - 2 101 117
          BC007858.2 - 1 - 2 70 4299832 118
          D49743.1 - 1 - 2 74 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010854.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001318.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AF218018.1,AK290584.1,BC007858.2,D49743.1,NM_002192.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene INHBA

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          ovarian follicle development
          hair follicle development
          hematopoietic progenitor cell differentiation
          regulation of transcription from RNA polymerase II promoter
          defense response
          cell cycle arrest
          cell surface receptor signaling pathway
          cell-cell signaling
          nervous system development
          negative regulation of cell proliferation
          male gonad development
          positive regulation of gene expression
          positive regulation of pathway-restricted SMAD protein phosphorylation
          striatal medium spiny neuron differentiation
          cell differentiation
          erythrocyte differentiation
          negative regulation of cell growth
          positive regulation of cellular protein metabolic process
          activin receptor signaling pathway
          endodermal cell differentiation
          growth
          negative regulation of phosphorylation
          odontogenesis
          response to drug
          hemoglobin biosynthetic process
          progesterone secretion
          regulation of MAPK cascade
          negative regulation of interferon-gamma biosynthetic process
          negative regulation of B cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of macrophage differentiation
          negative regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of follicle-stimulating hormone secretion
          positive regulation of follicle-stimulating hormone secretion
          negative regulation of follicle-stimulating hormone secretion
          mesodermal cell differentiation
          cell development
          palate development
          positive regulation of ovulation
          SMAD protein signal transduction
          eyelid development in camera-type eye
          cellular response to follicle-stimulating hormone stimulus
          cellular response to cholesterol
          GABAergic neuron differentiation
          extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          protein binding
          growth factor activity
          peptide hormone binding
          inhibin binding
          identical protein binding
          protein heterodimerization activity
          type II activin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07020342.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020342.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020342.hg.1 TC07001318.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07020338.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020338.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020338.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020338.hg.1 TC07001318.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07020343.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020343.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020343.hg.1 TC07001318.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07020340.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020340.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07020340.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07020340.hg.1 TC07001318.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177157.hg.1 TC0700010854.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700177160.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177160.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177160.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177160.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177160.hg.1 TC0700010854.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 ENST00000464515.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay TC0700010854.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 ENST00000442711.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177156.hg.1 TC0700010854.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700177159.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177159.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177159.hg.1 ENST00000242208.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0700177159.hg.1 NM_002192.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700177159.hg.1 TC0700010854.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline