Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04188276_m1
          See other IGF2 GE Assays ›
          Gene Symbol:
          IGF2
          Gene Name:
          insulin like growth factor 2
          Gene Aliases:
          C11orf43, GRDF, IGF-II, PP9974
          Chromosome Location:
          Chr.11: 2129112 - 2149603 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04188276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IGF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127598.2 NP_001121070.1 2 - 3 436 83
          GenBank mRNA BP233290.1 - 2 - 3 230 83
          CN277570.1 - 2 - 3 423 83
          HM481219.1 - 2 - 3 298 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC11001274.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ossification
          positive regulation of protein phosphorylation
          platelet degranulation
          glucose metabolic process
          regulation of gene expression by genetic imprinting
          regulation of transcription, DNA-templated
          multicellular organism development
          female pregnancy
          memory
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          response to radiation
          response to nutrient levels
          response to estradiol
          response to nicotine
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          wound healing
          positive regulation of activated T cell proliferation
          response to drug
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          cellular protein metabolic process
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of mitotic nuclear division
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of insulin receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell division
          positive regulation of protein kinase B signaling
          cellular response to mechanical stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of receptor activity
          positive regulation of glycogen (starch) synthase activity
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein binding
          growth factor activity
          receptor activator activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000300632.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000337883.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381319.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381389.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381392.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381395.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000381406.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000416167.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000418738.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000434045.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 ENST00000487262.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 NM_000612.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 NM_001007139.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 NM_001127598.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 NR_003512.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016114.hg.1 TC11001274.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0