| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000450.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000450.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000450.hg.1 |
TC6_cox_hap2000038.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000338.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000338.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000338.hg.1 |
TC6_mcf_hap5000025.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000433.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000433.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000433.hg.1 |
TC6_ssto_hap7000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000434.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000434.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000434.hg.1 |
TC6_ssto_hap7000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010338.hg.1 |
ENST00000428249.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002904.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002904.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002904.hg.1 |
TC06000340.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000440.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000440.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000440.hg.1 |
TC6_dbb_hap3000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000431.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000431.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000431.hg.1 |
TC6_mann_hap4000033.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000432.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000432.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000432.hg.1 |
TC6_ssto_hap7000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000449.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000449.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000449.hg.1 |
TC6_cox_hap2000038.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000439.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000439.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000439.hg.1 |
TC6_dbb_hap3000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000430.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000430.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000430.hg.1 |
TC6_mann_hap4000033.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000339.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000339.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000339.hg.1 |
TC6_mcf_hap5000025.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000435.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000435.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000435.hg.1 |
TC6_qbl_hap6000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000436.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000436.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000436.hg.1 |
TC6_qbl_hap6000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000437.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000437.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000437.hg.1 |
TC6_qbl_hap6000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
ENST00000376560.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
ENST00000498336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002903.hg.1 |
TC06000340.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000451.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000451.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000451.hg.1 |
TC6_cox_hap2000038.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000438.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000438.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000438.hg.1 |
TC6_dbb_hap3000031.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000429.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000429.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000429.hg.1 |
TC6_mann_hap4000033.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000340.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000340.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000340.hg.1 |
TC6_mcf_hap5000025.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
ENST00000376555.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
ENST00000376560.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
ENST00000498336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
NM_001077497.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002901.hg.1 |
TC06000340.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600150997.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150997.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150997.hg.1 |
TC0600007534.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150994.hg.1 |
TC0600007534.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010338.hg.1 |
ENST00000428249.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000491173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007534.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000376557.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000376560.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000470703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000481741.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
ENST00000498336.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
NM_025263.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150996.hg.1 |
TC0600007534.hg.1 |
Clariom D TC |