Search
Search

CST9L| Gene Symbol: CST9L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080610.2 | NP_542177.1 | 2 - 3 | 651 | 107 | |
| GenBank mRNA | AK312112.1 | - | 2 - 3 | 649 | 107 | |
| AY358615.1 | - | 2 - 3 | 594 | 107 | ||
| BC029656.1 | - | 2 - 3 | 651 | 5163974 | 107 | |
Set Membership: |
|