Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04230774_s1
          See other TCF21 GE Assays ›
          Gene Symbol:
          TCF21 TARID
          Gene Name:
          transcription factor 21
          TCF21 antisense RNA inducing promoter demethylation
          Gene Aliases:
          POD1, bHLHa23
          EYA4-AS1
          Chromosome Location:
          Chr.6: 133889121 - 133895537 on Build GRCh38
          Chromosome Location:
          Chr.6: 133502252 - 133889006 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04230774_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TCF21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003206.3 NP_003197.2 2 948 76
          GenBank mRNA AF035718.1 - 2 932 76
          AF047419.1 - 2 931 76
          BU688835.1 - 1 76
          W73859.1 - 2 321 76
          Gene Symbol: TARID
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA KF484511.1 - 1 76
          KF484512.1 - 1 76
          KF484513.1 - 1 76
          KF484514.1 - 1 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TCF21
          Click here to find siRNAs for Gene TARID

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          branching involved in ureteric bud morphogenesis
          morphogenesis of a branching structure
          kidney development
          vasculature development
          transcription from RNA polymerase II promoter
          sex determination
          branchiomeric skeletal muscle development
          epithelial cell differentiation
          glomerulus development
          positive regulation of transcription from RNA polymerase II promoter
          lung alveolus development
          spleen development
          embryonic digestive tract morphogenesis
          reproductive structure development
          gland development
          Sertoli cell differentiation
          palate development
          lung morphogenesis
          lung vasculature development
          bronchiole development
          diaphragm development
          respiratory system development
          negative regulation of androgen receptor signaling pathway
          metanephric mesenchymal cell differentiation
          metanephric glomerular capillary formation
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          histone deacetylase binding
          bHLH transcription factor binding
          protein dimerization activity
          androgen receptor binding
          E-box binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000419627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000451017.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000606544.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607573.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000607641.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000626104.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630119.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000630728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 ENST00000631362.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013220.hg.1 NR_109982.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227494 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline