Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04260487_m1
See other DTNB GE Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04260487_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
2,694.00
Your Price
Online offer:
2694.0
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001320934.1
NP_001307863.1
14 - 15
1725
68
NM_001320936.1
NP_001307865.1
15 - 16
1815
68
NM_021907.4
NP_068707.1
15 - 16
1815
68
NM_033147.3
NP_149159.2
15 - 16
1815
68
NM_033148.3
NP_149160.1
14 - 15
1725
68
NM_183360.2
NP_899204.1
15 - 16
1815
68
XM_011532671.1
XP_011530973.1
15 - 16
1750
68
XM_011532672.2
XP_011530974.1
15 - 16
1750
68
XM_011532674.1
XP_011530976.1
15 - 16
1750
68
XM_011532676.1
XP_011530978.1
14 - 15
1660
68
XM_011532677.2
XP_011530979.1
14 - 15
2206
68
XM_011532679.1
XP_011530981.1
15 - 16
1750
68
XM_011532680.2
XP_011530982.1
14 - 15
1671
68
XM_011532681.2
XP_011530983.1
14 - 15
1646
68
XM_017003532.1
XP_016859021.1
14 - 15
1660
68
XM_017003533.1
XP_016859022.1
14 - 15
1642
68
XM_017003536.1
XP_016859025.1
14 - 15
1642
68
XM_017003537.1
XP_016859026.1
11 - 12
2103
68
XM_017003539.1
XP_016859028.1
13 - 14
1517
68
XM_017003540.1
XP_016859029.1
13 - 14
1556
68
XM_017003541.1
XP_016859030.1
13 - 14
1540
68
XM_017003542.1
XP_016859031.1
13 - 14
1512
68
XM_017003543.1
XP_016859032.1
12 - 13
1406
68
XM_017003544.1
XP_016859033.1
13 - 14
1494
68
XM_017003545.1
XP_016859034.1
12 - 13
1388
68
GenBank mRNA
AF022728.1
-
15 - 16
1713
68
AF070567.1
-
9 - 10
957
68
AK094494.1
-
14 - 15
1533
68
BC049366.1
-
15 - 16
1806
4817552
68
Y12712.1
-
14 - 15
1645
68
Y15718.1
-
14 - 15
1552
68
Y15722.1
-
14 - 15
1552
68
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Homo sapiens
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene DTNB
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline