Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273773_m1
          See other LINC01235 GE Assays ›
          Gene Symbol:
          LINC01235
          Gene Name:
          long intergenic non-protein coding RNA 1235
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 13406380 - 13431329 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273773_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 4,565.00
          Your Price
          Online offer:
          4565.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01235
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033863.1 1 - 2 128 90
          GenBank mRNA AK123194.1 - 1 - 2 128 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA177332.1 - 1 - 2 123 Chr. 9 - 13408184 - 13431324 [-] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream multiple PDZ domain crumbs cell polarity complex component MPDZ HYC2,
          MUPP1
          -136.7kb Sense Chr. 9 - 13105707 - 13279692 on NCBI Build GRCh38
          Downstream uncharacterized LOC105375977 LOC105375977 9.4kb Anti-sense Chr. 9 - 13440735 - 13487507 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009561.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000915.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09012170.hg.1 NR_033863.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09012170.hg.1 TC09000915.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09012171.hg.1 NR_033863.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09012171.hg.1 TC09000915.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay TC0900009561.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 ENST00000605459.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900165469.hg.1 TC0900009561.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 ENST00000604724.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 NR_033863.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900165470.hg.1 TC0900009561.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline