| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000506222.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525746.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000525912.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000531046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
ENST00000533304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011569.hg.1 |
NR_038989.1 |
Refseq |