| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020822.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020822.hg.1 |
TC0X001552.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020823.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020823.hg.1 |
TC0X001552.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176905.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176905.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176905.hg.1 |
TC0X00011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00160926.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160926.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160926.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160926.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160926.hg.1 |
TC0X00008878.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00176906.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176906.hg.1 |
NR_109985.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176906.hg.1 |
TC0X00011241.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00160924.hg.1 |
TC0X00008878.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000433624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000452506.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
ENST00000541151.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008878.hg.1 |
NR_039991.1 |
Refseq |