Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274739_m1
          See other TMLHE-AS1 GE Assays ›
          Gene Symbol:
          TMLHE-AS1 LOC101927830
          Gene Name:
          TMLHE antisense RNA 1
          uncharacterized LOC101927830
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.X: 155466540 - 155494110 on Build GRCh38
          Chromosome Location:
          Chr.X: 155347137 - 155374174 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04274739_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TMLHE-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039991.1 3 - 4 469 156
          GenBank mRNA BC042086.1 - 3 - 4 469 156
          Gene Symbol: LOC101927830
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_109985.1 3 - 4 469 156
          GenBank mRNA BQ072670.1 - 3 - 4 488 156

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream trimethyllysine hydroxylase,
          epsilon
          TMLHE AUTSX6,
          BBOX2,
          TMLD,
          TMLH,
          TMLHED,
          XAP130
          Anti-sense Chr. X - 155489011 - 155612952 on NCBI Build GRCh38
          Downstream sprouty RTK signaling antagonist 3 SPRY3 spry-3 118.5kb Sense Chr. X - 155612586 - 155782459 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00008878.hg.1, TC0X00011241.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X001552.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P12039900 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P112380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8171033 - -
          Affymetrix Human Gene 1.0 ST Array 8176244 - -
          Affymetrix Human Gene 1.1 ST Array 8171032 - -
          Affymetrix Human Gene 1.1 ST Array 8176243 - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000452506.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000452506.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X020819.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X020819.hg.1 NR_039991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X020819.hg.1 TC0X001552.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 ENST00000444722.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00011241.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00160929.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00160929.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00160929.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00160929.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00160929.hg.1 TC0X00008878.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000433624.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000452506.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000452506.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 ENST00000541151.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00008878.hg.1 NR_039991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176899.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176899.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176899.hg.1 ENST00000412436.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00176899.hg.1 NR_109985.1 Refseq
          Affymetrix Human Clariom D Assay PSR0X00176899.hg.1 TC0X00011241.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1560797_s_at - -
          Human Genome U133 Plus 2.0 Array 1560797_PM_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560797_s_at2 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041791 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040618 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A041791 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040618 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1831764 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5960041 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_123848 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1831764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline