Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04275606_s1
          See other CFAP95-DT GE Assays ›
          Gene Symbol:
          CFAP95-DT
          Gene Name:
          CFAP95 divergent transcript
          Gene Aliases:
          C9orf135-AS1, C9orf135-DT
          Chromosome Location:
          Chr.9: 69819405 - 69820739 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04275606_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CFAP95-DT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038833.1 2 719 96
          GenBank mRNA BC039385.1 - 2 741 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB508203.1 - 1 198 Chr. 9 - 69819407 - 69819836 [-] on Build GRCh38 96
          GenBankmRNA AW015053.1 - 1 246 Chr. 9 - 69819401 - 69819890 [+] on Build GRCh38 96
          GenBankmRNA HY196629.1 - 1 210 Chr. 9 - 69819459 - 69819958 [+] on Build GRCh38 96
          GenBankmRNA HY221435.1 - 1 177 Chr. 9 - 69819463 - 69819808 [+] on Build GRCh38 96
          GenBankmRNA BF223409.1 - 1 241 Chr. 9 - 69819401 - 69819805 [+] on Build GRCh38 96
          GenBankmRNA HY198033.1 - 1 179 Chr. 9 - 69819461 - 69820023 [+] on Build GRCh38 96
          GenBankmRNA AI350410.1 - 1 203 Chr. 9 - 69819437 - 69819769 [+] on Build GRCh38 96
          GenBankmRNA AW117391.1 - 1 187 Chr. 9 - 69819456 - 69819908 [+] on Build GRCh38 96
          GenBankmRNA AI657094.1 - 1 232 Chr. 9 - 69819409 - 69819874 [+] on Build GRCh38 96
          GenBankmRNA AA431615.1 - 1 444 Chr. 9 - 69819603 - 69820082 [-] on Build GRCh38 96
          GenBankmRNA BU852367.1 - 1 92 Chr. 9 - 69819005 - 69819728 [-] on Build GRCh38 96
          GenBankmRNA AA431327.1 - 1 229 Chr. 9 - 69819413 - 69819833 [+] on Build GRCh38 96
          GenBankmRNA AI948877.1 - 1 255 Chr. 9 - 69819411 - 69819722 [+] on Build GRCh38 96
          GenBankmRNA HY224623.1 - 1 179 Chr. 9 - 69819461 - 69819910 [+] on Build GRCh38 96
          GenBankmRNA HY202151.1 - 1 179 Chr. 9 - 69819461 - 69819947 [+] on Build GRCh38 96
          GenBankmRNA HY192168.1 - 1 179 Chr. 9 - 69819461 - 69819915 [+] on Build GRCh38 96
          GenBankmRNA BE041843.1 - 1 245 Chr. 9 - 69819401 - 69819877 [+] on Build GRCh38 96
          GenBankmRNA HY244270.1 - 1 179 Chr. 9 - 69819461 - 69819948 [+] on Build GRCh38 96
          GenBankmRNA AI206930.1 - 1 232 Chr. 9 - 69819409 - 69819860 [+] on Build GRCh38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protein prenyltransferase alpha subunit repeat containing 1 PTAR1 -59.4kb Sense Chr. 9 - 69709525 - 69760011 on NCBI Build GRCh38
          Downstream cilia and flagella associated protein 95 CFAP95 C9orf135 550.0 Anti-sense Chr. 9 - 69820817 - 69906227 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900010355.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001195.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 512925 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P173862 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21131 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P173862 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P173862 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38739858 - -
          Agilent Human 1B Microarray G4111A A_32_P173862 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P38739858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 236071_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR09015316.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09015316.hg.1 NR_038833.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09015316.hg.1 TC09001195.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09015312.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09015312.hg.1 NR_038833.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09015312.hg.1 TC09001195.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 ENST00000453410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 ENST00000453410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170860.hg.1 TC0900010355.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000453410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000526458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000453410.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 ENST00000526458.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170861.hg.1 TC0900010355.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900170867.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170867.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170867.hg.1 ENST00000439418.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900170867.hg.1 NR_038833.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900170867.hg.1 TC0900010355.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 236071_PM_at - -
          Human Genome U133 Plus 2.0 Array 236071_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236071_at5 - -
          Human Genome U95 Set 76652_at - -
          Human Genome U95 Set 76652_PM_at - -
          Human Genome U133 Set 236071_PM_at - -
          Human Genome U133 Set 236071_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037552 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A037552 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 611688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline