Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331720_m1
          See other HNRNPKP3 GE Assays ›
          Gene Symbol:
          HNRNPKP3
          Gene Name:
          heterogeneous nuclear ribonucleoprotein K pseudogene 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 43261504 - 43269369 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331720_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HNRNPKP3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033868.1 1 - 2 105 87
          GenBank mRNA AK126814.1 - 1 - 2 105 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA139379.1 - 1 - 2 105 Chr. 11 - 43262750 - 43269369 [-] on Build GRCh38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100507205 LOC100507205 -1.0mb Sense Chr. 11 - 42187743 - 42253690 on NCBI Build GRCh38
          Downstream apoptosis inhibitor 5 API5 AAC-11,
          AAC11
          42.6kb Anti-sense Chr. 11 - 43311955 - 43344530 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010599.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001570.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183040 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439369 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439366 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183074 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439374 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183076 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439382 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781921 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439377 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782004 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07781977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07782030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16439378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28182993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28183082 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 214659 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 545667 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 102913 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 792193 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P714134 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21901 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61089 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3254590 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P714134 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02438122 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02438122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11019659.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019659.hg.1 NR_033868.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019659.hg.1 TC11001570.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019658.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11019658.hg.1 NR_033868.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11019658.hg.1 TC11001570.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100180333.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180333.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180333.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180333.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180333.hg.1 TC1100010599.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay TC1100010599.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 ENST00000534639.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180334.hg.1 TC1100010599.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 ENST00000511537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 ENST00000533565.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 NR_033868.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100180336.hg.1 TC1100010599.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045087 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045087 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline