Search
Search

HOXD1| Gene Symbol: HOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024501.3 | NP_078777.1 | 1 - 2 | 784 | 65 | |
| GenBank mRNA | AK314120.1 | - | 1 - 2 | 788 | 65 | |
| BC014477.1 | - | 1 - 2 | 777 | 65 | ||
Set Membership: |
|