| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
ENST00000064571.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009544.hg.1 |
NM_080617.5 |
Refseq |