Search
Search

TRIM69| Gene Symbol: TRIM69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301144.1 | NP_001288073.1 | 4 - 5 | 713 | 107 | |
| NM_080745.4 | NP_542783.2 | 4 - 5 | 705 | 107 | ||
| NM_182985.4 | NP_892030.3 | 5 - 6 | 1182 | 107 | ||
| GenBank mRNA | AF302088.1 | - | 4 - 5 | 561 | 107 | |
| AK226115.1 | - | 3 - 4 | 4969 | 107 | ||
| AK292252.1 | - | 5 - 6 | 1026 | 107 | ||
| AL360161.1 | - | 4 - 5 | 548 | 107 | ||
| AY305385.1 | - | 5 - 6 | 880 | 107 | ||
| BC024199.1 | - | 5 - 6 | 1000 | 107 | ||
| BC033314.1 | - | 4 - 5 | 501 | 5166812 | 107 | |
| BC047945.1 | - | 5 - 6 | 1019 | 5744224 | 107 | |
Set Membership: |
|