| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000507420.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400006692.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146395.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000512914.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146396.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000320912.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000382581.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
ENST00000617365.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_001272053.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_001272054.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
NM_033296.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400146397.hg.1 |
TC0400006692.hg.1 |
Clariom D TC |