| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023080.hg.1 |
ENST00000525544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023080.hg.1 |
ENST00000530719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023080.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023080.hg.1 |
TC11001925.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023083.hg.1 |
ENST00000525544.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023083.hg.1 |
ENST00000530719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023083.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023083.hg.1 |
TC11001925.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023082.hg.1 |
ENST00000530719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023082.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11023082.hg.1 |
TC11001925.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185858.hg.1 |
TC1100011202.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011202.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000525544.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185856.hg.1 |
TC1100011202.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000334821.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000524603.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000524632.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000528029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
ENST00000530719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
NM_001242631.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
NM_001282448.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185854.hg.1 |
TC1100011202.hg.1 |
Clariom D TC |