Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04402320_m1
          See other FLJ32255 GE Assays ›
          Gene Symbol:
          FLJ32255
          Gene Name:
          uncharacterized LOC643977
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 42985398 - 42993333 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04402320_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ32255
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104643.1 1 - 2 2492 123
          GenBank mRNA AK056817.1 - 1 - 2 2492 123
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AL578327.3 - 1 - 2 Chr. 5 - 42985444 - 42991417 [+] on Build GRCh38 123
          GenBankmRNA BE875232.1 - 128 Chr. - - [-] on Build 123
          GenBankmRNA HY285688.1 - 1 - 2 Chr. 5 - 42985398 - 42991046 [+] on Build GRCh38 123
          GenBankmRNA DB520017.1 - 1 - 2 276 Chr. 5 - 42985401 - 42991117 [-] on Build GRCh38 123
          GenBankmRNA CB306514.1 - 1 - 2 Chr. 5 - 42985398 - 42991104 [+] on Build GRCh38 123
          GenBankmRNA AA973343.1 - 1 - 2 Chr. 5 - 42985401 - 42990972 [+] on Build GRCh38 122
          GenBankmRNA HY223447.1 - 1 - 2 Chr. 5 - 42985401 - 42991030 [+] on Build GRCh38 123
          GenBankmRNA HY274641.1 - 1 - 2 Chr. 5 - 42985401 - 42991035 [+] on Build GRCh38 123
          GenBankmRNA AW958070.1 - 1 - 2 355 Chr. 5 - 42985429 - 42991196 [-] on Build GRCh38 123
          GenBankmRNA DB329338.1 - 1 - 2 Chr. 5 - 42985401 - 42991082 [+] on Build GRCh38 123
          GenBankmRNA AI278277.1 - 1 - 2 Chr. 5 - 42985401 - 42991015 [+] on Build GRCh38 123
          GenBankmRNA AI208978.1 - 1 - 2 Chr. 5 - 42985398 - 42990958 [+] on Build GRCh38 123
          GenBankmRNA AI807266.1 - 1 - 2 Chr. 5 - 42985398 - 42991152 [+] on Build GRCh38 123
          GenBankmRNA BX089745.1 - 1 - 2 344 Chr. 5 - 42985398 - 42991184 [-] on Build GRCh38 123
          GenBankmRNA HY226653.1 - 2 - 3 Chr. 5 - 42985401 - 42991110 [+] on Build GRCh38 123

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream selenoprotein P,
          plasma,
          1
          SEPP1 SELP,
          SEPP,
          SeP
          -173.5kb Sense Chr. 5 - 42799880 - 42811922 on NCBI Build GRCh38
          Downstream uncharacterized LOC648987 LOC648987 21.4kb Sense Chr. 5 - 43014729 - 43067439 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010625.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0500174155.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174155.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174155.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174157.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174157.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174157.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174156.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174156.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174156.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174159.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174159.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174159.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174160.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174160.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174160.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174161.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174161.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174161.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174154.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174154.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174154.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174162.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174162.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174162.hg.1 TC0500010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500174163.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174163.hg.1 NR_104643.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500174163.hg.1 TC0500010625.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline