| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100011251.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186385.hg.1 |
TC1100011251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000527453.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
ENST00000534178.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186386.hg.1 |
TC1100011251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100186384.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186384.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186384.hg.1 |
TC1100011251.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100186390.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186390.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186390.hg.1 |
ENST00000532454.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100186390.hg.1 |
NR_108085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100186390.hg.1 |
TC1100011251.hg.1 |
Clariom D TC |