Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04404153_g1
          See other LOC101929570 GE Assays ›
          Gene Symbol:
          LOC101929570
          Gene Name:
          uncharacterized LOC101929570
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 162768936 - 162797972 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04404153_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC101929570
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110258.2 3 - 4 255 109
          GenBank mRNA BC042876.1 - 3 - 4 255 109
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BG721990.1 - 3 - 4 260 Chr. 2 - 162768936 - 162795762 [+] on Build GRCh38 109
          GenBankmRNA AA430138.1 - 2 - 3 205 Chr. 2 - 162771929 - 162785254 [+] on Build GRCh38 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream potassium voltage-gated channel subfamily H member 7 KCNH7 ERG3,
          HERG3,
          Kv11.3
          Anti-sense Chr. 2 - 162371407 - 162838747 on NCBI Build GRCh38
          Downstream fidgetin,
          microtubule severing factor
          FIGN 819.8kb Anti-sense Chr. 2 - 163602611 - 163736155 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009832.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009832.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200168683.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200168683.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200168683.hg.1 ENST00000446838.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200168683.hg.1 NR_110258.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200168683.hg.1 TC0200009832.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline