| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009787.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009787.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009787.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009787.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000009787.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000166810.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000166810.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000166810.hg.1 |
ENST00000379256.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000166810.hg.1 |
NR_126062.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000166810.hg.1 |
TC1000009787.hg.1 |
Clariom D TC |