Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04416223_s1
          See other CATSPER2P1 GE Assays ›
          Gene Symbol:
          CATSPER2P1
          Gene Name:
          cation channel sperm associated 2 pseudogene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 43735934 - 43746298 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04416223_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CATSPER2P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002318.2 1 168 77
          GenBank mRNA BC047442.1 - 1 167 77
          BC066967.1 - 1 168 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB485515.1 - 1 168 Chr. 15 - 43745019 - 43746298 [-] on Build GRCh38 77
          GenBankmRNA HY177075.1 - 1 154 Chr. 15 - 43744494 - 43746284 [-] on Build GRCh38 77
          GenBankmRNA BG709135.1 - 1 173 Chr. 15 - 43736864 - 43746302 [-] on Build GRCh38 77
          GenBankmRNA BU602812.1 - 162 Chr. - - [-] on Build 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream stereocilin pseudogene 1 STRCP1 STRCP -27.8kb Sense Chr. 15 - 43699379 - 43718262 on NCBI Build GRCh38
          Downstream protein disulfide isomerase family A member 3 PDIA3 ER60,
          ERp57,
          ERp60,
          ERp61,
          GRP57,
          GRP58,
          HEL-S-269,
          HEL-S-93n,
          HsT17083,
          P58,
          PI-PLC
          94.0 Anti-sense Chr. 15 - 43746392 - 43772606 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500010861.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001284.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15012209.hg.1 ENST00000381680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15012209.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15012209.hg.1 TC15001284.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15012206.hg.1 ENST00000381680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15012206.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15012206.hg.1 TC15001284.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15012207.hg.1 ENST00000381680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15012207.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15012207.hg.1 TC15001284.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15012204.hg.1 ENST00000381680.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15012204.hg.1 NR_002318.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15012204.hg.1 TC15001284.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010861.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184466.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184469.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184469.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184469.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184469.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184469.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184468.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184468.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184468.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184468.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184468.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184460.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 ENST00000429276.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184465.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 ENST00000416302.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184471.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184459.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184459.hg.1 NR_002318.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500184459.hg.1 TC1500010861.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500184459.hg.1 ENST00000381680.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500184459.hg.1 NR_002318.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 188141 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline