Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04930436_g1
          See other RPPH1 GE Assays ›
          Gene Symbol:
          RPPH1 PARP2
          Gene Name:
          ribonuclease P RNA component H1
          poly(ADP-ribose) polymerase 2
          Gene Aliases:
          H1RNA, RPPH1-1
          ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
          Chromosome Location:
          Chr.14: 20343071 - 20343411 on Build GRCh38
          Chromosome Location:
          Chr.14: 20343041 - 20357904 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04930436_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPPH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002312.1 1 43 87
          GenBank mRNA CA413366.1 - 1 87
          DW429001.1 - 1 59 87
          X15624.1 - 1 42 87
          Gene Symbol: PARP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017020912.1 XP_016876401.1 1 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008586.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000882.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_017020912.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lagging strand elongation
          DNA repair
          base-excision repair
          protein ADP-ribosylation
          DNA ligation involved in DNA repair
          extrinsic apoptotic signaling pathway
          DNA binding
          DNA ligase (ATP) activity
          NAD+ ADP-ribosyltransferase activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420327 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3389394 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3747 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3746 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47736 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56436 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3389394 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3389394 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3420327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29610642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09154129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09154131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09154131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09154129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29610642 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154131 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154102 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154098 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09154129 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002312_at - -
          Affymetrix Human Gene 1.0 ST Array 7977510 - -
          Affymetrix Human Gene 1.0 ST Array 7977509 - -
          Affymetrix Human Gene 1.0 ST Array 7977508 - -
          Affymetrix Human Gene 1.1 ST Array 7977507 - -
          Affymetrix HT HG-U133+ PM Array Plate 1556684_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR14009565.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009565.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14009565.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14009565.hg.1 TC14000882.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008586.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161619.hg.1 TC1400008586.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161616.hg.1 TC1400008586.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 ENST00000516869.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 ENST00000554988.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 NR_002312.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400161618.hg.1 TC1400008586.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1556684_PM_at - -
          Human Genome U133 Plus 2.0 Array 1556684_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002312_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556684_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050551 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050551 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1704056 NR_002312.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1704056 CA413366.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1704056 NR_002312.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1704056 CA413366.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1704056 NR_002312.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1704056 CA413366.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1704056 NR_002312.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1704056 CA413366.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5700471 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_14345 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1704056 NR_002312.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1704056 CA413366.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1704056 NR_002312.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1704056 CA413366.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1704056 NR_002312.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1704056 CA413366.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline