Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04966975_g1
          See other IDH1 GE Assays ›
          Gene Symbol:
          IDH1
          Gene Name:
          isocitrate dehydrogenase (NADP(+)) 1, cytosolic
          Gene Aliases:
          HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD
          Chromosome Location:
          Chr.2: 208236227 - 208255143 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04966975_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IDH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282386.1 NP_001269315.1 7 - 8 1057 88
          NM_001282387.1 NP_001269316.1 7 - 8 1214 88
          NM_005896.3 NP_005887.2 7 - 8 1146 88
          GenBank mRNA AF020038.1 - 7 - 8 1100 88
          AF113917.1 - 7 - 8 1037 88
          AK302105.1 - 5 - 6 592 88
          AK312242.1 - 7 - 8 1117 88
          AL136702.1 - 7 - 8 1078 88
          BC012846.1 - 7 - 8 1097 3880331 88
          BC021046.1 - 7 - 8 1043 88
          BC093020.1 - 7 - 8 1075 30528912 88
          BX537411.1 - 7 - 8 88
          CR533522.1 - 5 - 6 851 88
          CR536499.1 - 5 - 6 851 88
          CR541695.1 - 5 - 6 851 88
          DQ403082.1 - 5 - 6 764 88
          EU794623.1 - 7 - 8 1085 88
          FJ224342.1 - 7 - 8 88

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IDH1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glyoxylate cycle
          tricarboxylic acid cycle
          isocitrate metabolic process
          2-oxoglutarate metabolic process
          NADPH regeneration
          glutathione metabolic process
          response to oxidative stress
          female gonad development
          response to steroid hormone
          regulation of phospholipid catabolic process
          regulation of phospholipid biosynthetic process
          cell-cell adhesion
          magnesium ion binding
          isocitrate dehydrogenase (NADP+) activity
          receptor binding
          protein homodimerization activity
          NADP binding
          NAD binding
          (R)-2-hydroxyglutarate dehydrogenase activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000345146.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415282.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000415913.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000417583.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000446179.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000451391.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000462386.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000481557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 ENST00000484575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282386.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_001282387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200015578.hg.1 NM_005896.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0