| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000297537.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
ENST00000475831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013061.hg.1 |
NM_001098834.2 |
Refseq |