Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04996927_m1
          See other TRIM32 GE Assays ›
          Gene Symbol:
          TRIM32
          Gene Name:
          tripartite motif containing 32
          Gene Aliases:
          BBS11, HT2A, LGMD2H, TATIP
          Chromosome Location:
          Chr.9: 116687302 - 116701300 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04996927_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIM32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017014486.1 XP_016869975.1 1 - 2 81 97

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_017014486.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          tissue homeostasis
          actin ubiquitination
          response to UV
          protein ubiquitination
          positive regulation of cell growth
          positive regulation of cell migration
          regulation of type I interferon production
          negative regulation of viral transcription
          response to tumor necrosis factor
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of protein catabolic process
          positive regulation of cell cycle
          positive regulation of proteolysis
          muscle cell cellular homeostasis
          negative regulation of fibroblast proliferation
          positive regulation of neurogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of striated muscle cell differentiation
          axon development
          negative regulation of viral release from host cell
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of interleukin-17-mediated signaling pathway
          positive regulation of chemokine (C-C motif) ligand 20 production
          positive regulation of cell motility
          transcription coactivator activity
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          myosin binding
          Tat protein binding
          translation initiation factor binding
          ubiquitin binding
          protein self-association
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000373983.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000411410.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 ENST00000450136.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_001099679.1 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008574.hg.1 NM_012210.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline