Search
Search

ARMCX1| Gene Symbol: ARMCX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016608.1 | NP_057692.1 | 4 | 694 | 86 | |
| GenBank mRNA | AB039670.1 | - | 4 | 694 | 86 | |
| AF248963.1 | - | 4 | 627 | 86 | ||
| AK074785.1 | - | 4 | 644 | 86 | ||
| AK222578.1 | - | 4 | 671 | 86 | ||
| BC002691.2 | - | 4 | 633 | 3605690 | 86 | |
Set Membership: |
|