Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05019740_g1
          See other S100A8 GE Assays ›
          Gene Symbol:
          S100A8
          Gene Name:
          S100 calcium binding protein A8
          Gene Aliases:
          60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
          Chromosome Location:
          Chr.1: 153390032 - 153422583 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05019740_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100A8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001319196.1 NP_001306125.1 2 - 3 325 89
          NM_001319197.1 NP_001306126.1 2 - 3 322 89
          NM_001319198.1 NP_001306127.1 2 - 3 226 89
          NM_001319201.1 NP_001306130.1 2 - 3 280 89
          NM_002964.4 NP_002955.2 2 - 3 308 89
          GenBank mRNA AA401538.1 - 2 - 3 223 89
          AA903259.1 - 1 - 2 89
          AI126134.1 - 1 - 2 89
          AK291328.1 - 2 - 3 193 89
          BC005928.2 - 1 - 2 160 4246359 89
          BP209293.1 - 2 - 3 210 89
          BP226540.1 - 2 - 3 208 89
          BT007378.1 - 1 - 2 138 89
          BU935536.1 - 2 - 3 310 89
          CR407674.1 - 1 - 2 138 89
          X06234.1 - 2 - 3 194 89
          Y00278.1 - 2 - 3 189 89

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100A8

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production
          leukocyte migration involved in inflammatory response
          acute inflammatory response
          chronic inflammatory response
          positive regulation of peptide secretion
          autophagy
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          response to zinc ion
          astrocyte development
          peptidyl-cysteine S-nitrosylation
          positive regulation of cell growth
          neutrophil chemotaxis
          sequestering of zinc ion
          response to lipopolysaccharide
          chemokine production
          wound healing
          defense response to bacterium
          innate immune response
          response to ethanol
          positive regulation of inflammatory response
          defense response to fungus
          positive regulation of NF-kappaB transcription factor activity
          regulation of cytoskeleton organization
          neutrophil aggregation
          positive regulation of intrinsic apoptotic signaling pathway
          calcium ion binding
          protein binding
          microtubule binding
          zinc ion binding
          Toll-like receptor 4 binding
          arachidonic acid binding
          RAGE receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368732.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000368733.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 ENST00000477801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015858.hg.1 NM_002964.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline