Search
Search
| Gene Symbol: BRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113182.2 | NP_001106653.1 | 2 | 1187 | 92 | |
| NM_001199455.1 | NP_001186384.1 | 1 | 877 | 92 | ||
| NM_001291986.1 | NP_001278915.1 | 2 | 1187 | 92 | ||
| NM_005104.3 | NP_005095.1 | 2 | 1274 | 92 | ||
| GenBank mRNA | AK056504.1 | - | 2 | 1145 | 92 | |
| AL832722.1 | - | 2 | 1122 | 92 | ||
| BC000477.2 | - | 1 | 246 | 92 | ||
| BX648109.1 | - | 2 | 1188 | 92 | ||
| D42040.1 | - | 2 | 1274 | 92 | ||
| M80613.1 | - | 1 | 609 | 92 | ||
| X62083.1 | - | 1 | 609 | 92 | ||
Set Membership: |
|