Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05042292_g1
          See other FEN1 GE Assays ›
          Gene Symbol:
          FEN1
          Gene Name:
          flap structure-specific endonuclease 1
          Gene Aliases:
          FEN-1, MF1, RAD2
          Chromosome Location:
          Chr.11: 61792637 - 61797244 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05042292_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FEN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004111.5 NP_004102.1 2 2138 92
          GenBank mRNA BC000323.2 - 2 1854 2821792 92
          BM450370.1 - 1 332 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FEN1

          Panther Classification:

          Molecular Function -

          exodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          double-strand break repair via homologous recombination
          DNA replication
          DNA repair
          base-excision repair
          double-strand break repair
          memory
          UV protection
          DNA replication, removal of RNA primer
          RNA phosphodiester bond hydrolysis, endonucleolytic
          magnesium ion binding
          DNA binding
          damaged DNA binding
          double-stranded DNA binding
          endonuclease activity
          RNA-DNA hybrid ribonuclease activity
          exonuclease activity
          protein binding
          double-stranded DNA exodeoxyribonuclease activity
          5'-3' exonuclease activity
          5'-flap endonuclease activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000535307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000535307.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000305885.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000535307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000535307.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 ENST00000535723.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq
          Affymetrix Human Clariom D Assay TC1100013021.hg.1 NM_004111.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline