Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05042293_s1
          See other CCND1 GE Assays ›
          Gene Symbol:
          CCND1
          Gene Name:
          cyclin D1
          Gene Aliases:
          BCL1, D11S287E, PRAD1, U21B31
          Chromosome Location:
          Chr.11: 69641105 - 69654474 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05042293_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCND1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053056.2 NP_444284.1 4 904 91
          GenBank mRNA AK299044.1 - 3 451 91
          AK313136.1 - 4 853 91
          BC000076.2 - 4 838 3508088 91
          BC001501.2 - 4 838 3507598 91
          BC014078.2 - 4 838 4124540 91
          BC023620.2 - 4 831 4650919 91
          BC025302.1 - 4 840 5457963 91
          BT019844.1 - 4 695 91
          BT019845.1 - 4 695 91
          CR536538.1 - 4 695 91
          CR542099.1 - 4 695 91
          M64349.1 - 4 839 91
          M73554.1 - 4 836 91
          M74092.1 - 4 844 91
          X59798.1 - 4 842 91

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCND1

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          re-entry into mitotic cell cycle
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          protein phosphorylation
          cellular response to DNA damage stimulus
          lactation
          response to iron ion
          response to X-ray
          response to organonitrogen compound
          positive regulation of G2/M transition of mitotic cell cycle
          negative regulation of Wnt signaling pathway
          negative regulation of epithelial cell differentiation
          endoplasmic reticulum unfolded protein response
          mitotic G1 DNA damage checkpoint
          response to magnesium ion
          response to estradiol
          response to vitamin E
          Leydig cell differentiation
          mammary gland epithelial cell proliferation
          positive regulation of mammary gland epithelial cell proliferation
          response to drug
          response to estrogen
          response to leptin
          fat cell differentiation
          response to ethanol
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell cycle
          cell division
          response to corticosterone
          response to calcium ion
          canonical Wnt signaling pathway
          mammary gland alveolus development
          response to UV-A
          negative regulation of cell cycle arrest
          liver regeneration
          transcription corepressor activity
          protein kinase activity
          protein binding
          transcription factor binding
          cyclin-dependent protein serine/threonine kinase regulator activity
          enzyme binding
          protein kinase binding
          protein complex binding
          histone deacetylase binding
          proline-rich region binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000227507.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000535993.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000536559.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000539241.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000542367.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 ENST00000545484.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq
          Affymetrix Human Clariom D Assay TC1100008236.hg.1 NM_053056.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline