Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05045233_s1
          See other LINC00491 GE Assays ›
          Gene Symbol:
          LINC00491
          Gene Name:
          long intergenic non-protein coding RNA 491
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 102608492 - 102671464 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05045233_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00491
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_103753.1 3 469 71
          NR_103754.1 4 454 71
          NR_103755.1 3 391 71
          NR_103756.1 2 320 71
          GenBank mRNA AK092078.1 - 2 611 71
          BG777293.1 - 2 271 71
          CN270888.1 - 4 297 71
          DA458115.1 - 3 288 71
          DA458823.1 - 3 391 71

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000505527.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000508339.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000510145.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 ENST00000513815.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103753.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103754.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103755.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011598.hg.1 NR_103756.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline