| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000436054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451152.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
ENST00000451596.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110952.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007556.hg.1 |
NR_110953.1 |
Refseq |