Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05050768_sH
          See other GREM1 GE Assays ›
          Gene Symbol:
          GREM1
          Gene Name:
          gremlin 1, DAN family BMP antagonist
          Gene Aliases:
          C15DUPq, CKTSF1B1, CRAC1, CRCS4, DAND2, DRM, DUP15q, GREMLIN, HMPS, HMPS1, IHG-2, MPSH, PIG2
          Chromosome Location:
          Chr.15: 32717940 - 32734669 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05050768_sH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GREM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013372.6 NP_037504.1 2 358 108
          XM_005254301.2 XP_005254358.1 2 422 108
          XM_017022077.1 XP_016877566.1 2 404 108
          GenBank mRNA AF045800.1 - 1 199 108
          AF110137.2 - 2 261 108
          AF154054.1 - 2 328 108
          AK312265.1 - 2 359 108
          AY232290.1 - 2 334 108
          BC069525.1 - 1 199 7262108 108
          BC093778.1 - 2 221 7939623 108
          BC101611.1 - 2 221 8069117 108

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GREM1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          branching involved in ureteric bud morphogenesis
          cell migration involved in sprouting angiogenesis
          positive regulation of receptor internalization
          positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          apoptotic process
          signal transduction
          activation of transmembrane receptor protein tyrosine kinase activity
          cell-cell signaling
          positive regulation of cell proliferation
          proximal/distal pattern formation
          regulation of epithelial to mesenchymal transition
          collagen fibril organization
          negative regulation of cell growth
          embryonic limb morphogenesis
          negative regulation of bone mineralization
          negative regulation of BMP signaling pathway
          negative regulation of chondrocyte differentiation
          regulation of stress-activated MAPK cascade
          negative regulation of osteoblast proliferation
          positive regulation of NF-kappaB import into nucleus
          negative regulation of apoptotic process
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of bone remodeling
          determination of dorsal identity
          positive regulation of NF-kappaB transcription factor activity
          regulation of focal adhesion assembly
          positive regulation of telomerase activity
          limb development
          negative regulation of pathway-restricted SMAD protein phosphorylation
          ureteric bud formation
          positive regulation of protein tyrosine kinase activity
          signal transduction by p53 class mediator
          negative regulation of monocyte chemotaxis
          negative regulation of canonical Wnt signaling pathway
          positive regulation of branching involved in ureteric bud morphogenesis
          negative regulation of branching involved in ureteric bud morphogenesis
          negative regulation of osteoclast proliferation
          positive regulation of peptidyl-tyrosine autophosphorylation
          negative regulation of bone trabecula formation
          negative regulation of bone mineralization involved in bone maturation
          positive regulation of receptor activity
          positive regulation of cardiac muscle cell differentiation
          cytokine activity
          protein binding
          morphogen activity
          transmembrane receptor protein tyrosine kinase activator activity
          BMP binding
          vascular endothelial growth factor receptor 2 binding
          receptor agonist activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560677.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000560830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 ENST00000622074.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191322.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_001191323.1 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq
          Affymetrix Human Clariom D Assay TC1500006788.hg.1 NM_013372.6 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline